ENA061957556 Enamine

2-[4-(3,5-dimethyl-1H-pyrazol-1-yl)phenyl]acetonitrile

  • MDL number MFCD07329987
  • Cas Number : 956986-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14063
Availability details for
Quantity
  • Availability
  • Add to Cart