ENA061957561 Enamine

3-amino-4-chloro-N-(3-morpholin-4-ylpropyl)benzenesulfonamide

  • MDL number MFCD07330153
  • Cas Number : 848658-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14068

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14068
Availability details for
Quantity
  • Availability
  • Add to Cart