ENA061957565 Enamine

6-[(methylsulfonyl)amino]hexanoic acid

  • MDL number MFCD01056017
  • Cas Number : 105612-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14072

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14072
Availability details for
Quantity
  • Availability
  • Add to Cart