ENA061957574 Enamine

2-[2-(2-oxopyrrolidin-1-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD07329997
  • Cas Number : 848658-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14081
Availability details for
Quantity
  • Availability
  • Add to Cart