ENA061957582 Enamine

ethyl (2E)-4-chloro-3-hydroxy-2-(4-methyl-1,3-thiazol-2-yl)but-2-enoate

  • MDL number MFCD07329999
  • Cas Number : 848658-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14089

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14089
Availability details for
Quantity
  • Availability
  • Add to Cart