ENA061957609 Enamine

2-[1-(4-chlorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]acetic acid

  • MDL number MFCD00866126
  • Cas Number : 42779-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14116
Availability details for
Quantity
  • Availability
  • Add to Cart