ENA061957612 Enamine

3-(1,3-benzothiazol-2-yl)pyrazine-2-carboxylic acid

  • MDL number MFCD03469002
  • Cas Number : 100283-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-191
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14119
Availability details for
Quantity
  • Availability
  • Add to Cart