ENA061957614 Enamine

3-(2-mercapto-4-oxoquinazolin-3(4H)-yl)propanoic acid

  • MDL number MFCD03030126
  • Cas Number : 451465-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14121

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14121
Availability details for
Quantity
  • Availability
  • Add to Cart