ENA061957620 Enamine

2-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzoic acid

  • MDL number MFCD07330271
  • Cas Number : 848316-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14127
Availability details for
Quantity
  • Availability
  • Add to Cart