ENA061957621 Enamine

morpholine-4-carbothiohydrazide

  • MDL number MFCD00588818
  • Cas Number : 6499-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14128

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14128
Availability details for
Quantity
  • Availability
  • Add to Cart