ENA061957638 Enamine

1,5-diamino-4-(1,3-benzothiazol-2-yl)-2,3-dihydro-1H-pyrrol-3-one

  • MDL number MFCD03653127
  • Cas Number : 610278-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14009
Availability details for
Quantity
  • Availability
  • Add to Cart