ENA061957640 Enamine

(3-fluoro-4-methoxyphenyl)acetonitrile

  • MDL number MFCD00040889
  • Cas Number : 404-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33...35
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14147

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-14147
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart