ENA061957642 Enamine

5-(chloromethyl)-3-methyl-1-phenyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD07329919
  • Cas Number : 698367-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-130
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14149

Safety Information

signalword Danger
pcodes P260,P261,P264,P271,P280,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-14149
pictograms GHS05,GHS07
hstatements H314,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart