ENA061957676 Enamine

2-(4-fluorophenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD07339052
  • Cas Number : 863668-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14183
Availability details for
Quantity
  • Availability
  • Add to Cart