ENA061957688 Enamine

4-(3,4-dimethylphenyl)-4-oxobutanoic acid

  • MDL number MFCD01320027
  • Cas Number : 51036-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14195
Availability details for
Quantity
  • Availability
  • Add to Cart