ENA065085406 Enamine

2-[(3,4-difluorophenyl)sulfanyl]acetic acid

  • MDL number MFCD07339314
  • Cas Number : 867311-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14239
Availability details for
Quantity
  • Availability
  • Add to Cart