ENA065085412 Enamine

1-(2-chloro-4,5-dimethoxyphenyl)ethanone

  • MDL number MFCD07339247
  • Cas Number : 161199-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14297

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14297
Availability details for
Quantity
  • Availability
  • Add to Cart