ENA065085413 Enamine

4-ethoxy-5-methoxy-2-nitrobenzoic acid

  • MDL number MFCD07339246
  • Cas Number : 103095-48-3
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-175
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14299
Availability details for
Quantity
  • Availability
  • Add to Cart