ENA065085415 Enamine

[2,5-dimethyl-1-(4-methylphenyl)-1H-pyrrol-3-yl]acetic acid

  • MDL number MFCD01688628
  • Cas Number : 42779-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50...52
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14308

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14308
Availability details for
Quantity
  • Availability
  • Add to Cart