ENA065085416 Enamine

2-[2-cyclopropyl-6-(methoxycarbonyl)-4-oxo-3H,4H-thieno[2,3-d]pyrimidin-5-yl]acetic acid

  • MDL number MFCD07339249
  • Cas Number : 863763-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14310
Availability details for
Quantity
  • Availability
  • Add to Cart