ENA065085425 Enamine

3-phenyl-4H-1,2,4-triazol-4-amine

  • MDL number MFCD07339278
  • Cas Number : 38345-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-100
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14325
Availability details for
Quantity
  • Availability
  • Add to Cart