ENA065085431 Enamine

ethyl 3-amino-5-(3,4-dimethoxyphenyl)thiophene-2-carboxylate

  • MDL number MFCD07339353
  • Cas Number : 872319-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14338
Availability details for
Quantity
  • Availability
  • Add to Cart