ENA065085435 Enamine

3,6-dimethyl-4-oxo-3H,4H-furo[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD07339362
  • Cas Number : 872319-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14346
Availability details for
Quantity
  • Availability
  • Add to Cart