ENA065085436 Enamine

2-{[(1-phenyl-1H-tetrazol-5-yl)methyl]thio}benzoic acid

  • MDL number MFCD07339364
  • Cas Number : 872319-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14348
Availability details for
Quantity
  • Availability
  • Add to Cart