ENA065085439 Enamine

1-butyl-2-(2-methoxyphenyl)-6-oxopiperidine-3-carboxylic acid

  • MDL number MFCD07339367
  • Cas Number : 872319-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 288...290
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14351

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14351
Availability details for
Quantity
  • Availability
  • Add to Cart