ENA065085441 Enamine

2-(3-cyclopropyl-1H-1,2,4-triazol-5-yl)aniline

  • MDL number MFCD07339539
  • Cas Number : 867329-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14360
Availability details for
Quantity
  • Availability
  • Add to Cart