ENA065085458 Enamine

3-(1-phenylethoxy)propan-1-amine

  • MDL number MFCD07339538
  • Cas Number : 867329-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250...254
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14387

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14387
Availability details for
Quantity
  • Availability
  • Add to Cart