ENA065085464 Enamine

1-[2-(4-hydroxyphenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one

  • MDL number MFCD07339544
  • Cas Number : 867330-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-134
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14401
Availability details for
Quantity
  • Availability
  • Add to Cart