ENA065085477 Enamine

2-bromo-4-(chloromethyl)-1-methoxybenzene

  • MDL number MFCD02856712
  • Cas Number : 701-94-0
  • Molecular Weight : 235.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 45-51
Iupac Name 2-bromo-4-(chloromethyl)-1-methoxybenzene
Inchi Code 1S/C8H8BrClO/c1-11-8-3-2-6(5-10)4-7(8)9/h2-4H,5H2,1H3
Inchi Key LGROUIFCRUBNIJ-UHFFFAOYSA-N

Description

Melting Point 45-51
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14422
Availability details for
Quantity
  • Availability
  • Add to Cart