ENA065085482 Enamine

2-[(pyrrolidin-1-yl)methyl]benzonitrile

  • MDL number MFCD07339548
  • Cas Number : 135277-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14435
Availability details for
Quantity
  • Availability
  • Add to Cart