ENA065085484 Enamine

1-(1-chloroethyl)-3-nitrobenzene

  • MDL number MFCD07343852
  • Cas Number : 34586-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33...35
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14438

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14438
Availability details for
Quantity
  • Availability
  • Add to Cart