ENA065085486 Enamine

2-(1-methyl-1H-pyrrol-2-yl)azepane

  • MDL number MFCD07339547
  • Cas Number : 867330-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14445
Availability details for
Quantity
  • Availability
  • Add to Cart