ENA065085487 Enamine

5-(chloroacetyl)-1-methyl-1,3-dihydro-2H-indol-2-one

  • MDL number MFCD07366366
  • Cas Number : 118306-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14447
Availability details for
Quantity
  • Availability
  • Add to Cart