ENA065085488 Enamine

2-(chloromethyl)-1-(4-methoxyphenyl)-1H-benzimidazole

  • MDL number MFCD07344336
  • Cas Number : 869716-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14449

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14449
Availability details for
Quantity
  • Availability
  • Add to Cart