ENA065085497 Enamine

1-[4-(bromoacetyl)phenyl]pyrrolidin-2-one

  • MDL number MFCD07345533
  • Cas Number : 870761-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14466

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-14466
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart