ENA065085499 Enamine

ethyl (4-cyclohexyl-5-mercapto-4H-1,2,4-triazol-3-yl)acetate

  • MDL number MFCD07339559
  • Cas Number : 867330-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14470

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14470
Availability details for
Quantity
  • Availability
  • Add to Cart