ENA065085502 Enamine

2-amino-N-(2,2,2-trifluoroethyl)benzamide

  • MDL number MFCD07343992
  • Cas Number : 869629-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-130
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14484
Availability details for
Quantity
  • Availability
  • Add to Cart