ENA065085504 Enamine

2-[6-(methoxycarbonyl)-2-methyl-4-oxo-3H,4H-thieno[2,3-d]pyrimidin-5-yl]acetic acid

  • MDL number MFCD07343865
  • Cas Number : 868238-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14488
Availability details for
Quantity
  • Availability
  • Add to Cart