ENA065085508 Enamine

7-(chloromethyl)-2-ethyl-5H-[1,3,4]thiadiazolo[3,2-a]pyrimidin-5-one

  • MDL number MFCD07343829
  • Cas Number : 869634-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97...99
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14494

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14494
Availability details for
Quantity
  • Availability
  • Add to Cart