ENA065085523 Enamine

2-[(1,3-benzothiazol-2-yl)amino]propanoic acid

  • MDL number MFCD07343835
  • Cas Number : 1397002-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14524
Availability details for
Quantity
  • Availability
  • Add to Cart