ENA065085524 Enamine

N-(sec-butyl)-1-methylhydrazinecarbothioamide

  • MDL number MFCD07343885

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14525

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14525
Availability details for
Quantity
  • Availability
  • Add to Cart