ENA065085525 Enamine

N-isobutyl-1-methylhydrazinecarbothioamide

  • MDL number MFCD07343836
  • Cas Number : 858481-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265...269
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14526

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14526
Availability details for
Quantity
  • Availability
  • Add to Cart