ENA065085526 Enamine

1-(4-methylphenyl)-2-(morpholin-4-yl)ethan-1-amine

  • MDL number MFCD07343838
  • Cas Number : 869634-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-37
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14527
Availability details for
Quantity
  • Availability
  • Add to Cart