ENA065085530 Enamine

2-[5-(chlorosulfonyl)thiophen-2-yl]acetic acid

  • MDL number MFCD07343895
  • Cas Number : 869709-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14543
Availability details for
Quantity
  • Availability
  • Add to Cart