ENA065085538 Enamine

6-amino-4-methyl-1,2-dihydropyridin-2-one

  • MDL number MFCD07344329
  • Cas Number : 51564-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247-248
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14559
Availability details for
Quantity
  • Availability
  • Add to Cart