ENA065085540 Enamine

1,1-dioxo-1lambda6-thiolane-3-carbohydrazide

  • MDL number MFCD07344331
  • Cas Number : 17236-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-107
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14561
Availability details for
Quantity
  • Availability
  • Add to Cart