ENA065085557 Enamine

3-(3-methyl-4-oxo-3,4-dihydroquinazolin-2-yl)propanoic acid

  • MDL number MFCD07344326
  • Cas Number : 869716-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14586
Availability details for
Quantity
  • Availability
  • Add to Cart