ENA065085565 Enamine

5-(1,1-dioxidotetrahydrothien-3-yl)pyrimidine-2,4,6(1H,3H,5H)-trione

  • MDL number MFCD07345586
  • Cas Number : 92259-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14602

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14602
Availability details for
Quantity
  • Availability
  • Add to Cart