ENA065085569 Enamine

5-(1,1-dioxidotetrahydrothien-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07345582
  • Cas Number : 869464-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14611

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14611
Availability details for
Quantity
  • Availability
  • Add to Cart