ENA065085578 Enamine

1-(thiophen-2-yl)-4,5-dihydro-1H-1,2,3,4-tetrazol-5-one

  • MDL number MFCD07345592
  • Cas Number : 141946-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14623
Availability details for
Quantity
  • Availability
  • Add to Cart